Mouse recombinant IL1 beta (from E. coli)
Fournisseur: ProSci Inc.
91-032
PRSI91-032EA 852 EUR InStock
PRSI91-032
Mouse recombinant IL1 beta (from E. coli)
Protéines et Peptides
Interleukin-1 (IL-1) designates two proteins, IL-1 alpha and IL-1 beta , which are the products of distinct genes, but recognize the same cell surface receptors. IL-1 alpha and IL-1 beta are structurally related polypeptides that show approximately 25% homology at the amino acid level. Both proteins are produced by a wide variety of cells in response to stimuli such as those produced by inflammatory agents, infections, or microbial endotoxins. The proteins are synthesized as 31 kDa precursors that are subsequently cleaved into proteins with molecular weights of approximately 17.5 kDa. The specific protease responsible for the processing of IL-1 beta , designated interleukin 1 beta -converting enzyme (ICE), has been described. Mature human and mouse IL-1 beta share approximately 75% amino acid sequence identity and human IL-1 beta has been found to be active on murine cell lines.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité