Mouse recombinant Growth hormone (from E. coli)
Fournisseur: ProSci Inc.
91-038
PRSI91-038EA 654 EUR InStock
PRSI91-038
Mouse recombinant Growth hormone (from E. coli)
Protéines et Peptides
Somatotropin(GH) is a member of the somatotropin/prolactin family of hormones which play an important role in growth control. Its major role in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1. GH stimulates both the differentiation and proliferation of myoblasts. It also stimulates amino acid uptake and protein synthesis in muscle and other tissues.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité