Human recombinant IL31 (from E. coli)
Fournisseur: ProSci Inc.
91-042
PRSI91-042EA 852 EUR InStock
PRSI91-042
Human recombinant IL31 (from E. coli)
Protéines et Peptides
Human Interleukin 31 (IL-31) is a cytokine containing a four-helix bundle structure. It shares several structural and functional characteristics with IL-6, Oncostatin M, LIF, and Cardiotrophin-1. Human IL-31 cDNA encodes a 164 amino acid precursor that contains a 23 amino acid signal peptide and a 141 amino acid mature protein. Human and mouse IL-31 share 24% sequence identity in the mature region. IL-31 is mainly associated with activated T cells and is preferentially expressed by type 2 helper T cells (Th2). IL-31 signals via a heterodimeric receptor complex composed of a gp130 related molecule termed IL-31RA (also GPL and GLMR) and an Oncostatin M receptor (OSM R beta ). The IL-31 receptor is constitutively expressed by keratinocytes and upregulated by IFN gamma on monocytes. GPL/OSMR signaling is a strong activator of STAT3 and STAT5, and can also activate STAT1, Jak1, and Jak2 signaling pathways. IL-31 regulated immune responses have been implicated in skin physiology and inflammatory skin diseases. Studies have shown that IL31 induces severe pruritis (itching) and dermatitis in transgenic mice.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité