Human Recombinant CCL8 (from E. coli)
Fournisseur: ProSci Inc.
91-050
PRSI91-050EA 852 EUR InStock
PRSI91-050
Human Recombinant CCL8 (from E. coli)
Protéines et Peptides
Human Chemokine (C-C Motif) Ligand 8 (CCL8) is produced by human MG63 osteosarcoma cells. CCL8 shares 62% and 58% amino acid sequence identity with MCP-1 and MCP-3, respectively. All three MCP proteins are monocyte chemoattractants. CCL8 is chemotactic for and activates many different immune cells, including mast cells, eosinophils and basophils, which are implicated in allergic response, and monocytes, T cells, and NK cells that are involved in the inflammatory response. CCL8 elicits its effects by binding to several different cell surface receptors including CCR1, CCR2B and CCR5.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité