Human Recombinant CCL27 (from E. coli)
Fournisseur: ProSci Inc.
91-052
PRSI91-052EA 655 EUR InStock
PRSI91-052
Human Recombinant CCL27 (from E. coli)
Protéines et Peptides
Human Chemokine (C-C Motif) Ligand 27 (CCL27) is a small cytokine that is a member of the CC chemokine family; it is expressed in numerous tissues, including gonads, thymus, placenta and skin. CCL27 elicits its chemotactic effects by binding to the chemokine receptor CCR10. Predominantly expressed in the skin, CCL27 is associated with T cell-mediated inflammation of the skin. Human and Mouse CCL27 share 84% sequence identity in the mature form.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité