Human recombinant IL1 alpha (from E. coli)
Fournisseur: ProSci Inc.
91-056
PRSI91-056EA 852 EUR InStock
PRSI91-056
Human recombinant IL1 alpha (from E. coli)
Protéines et Peptides
Interleukin-1 alpha (IL1 alpha) is a cytokine member of the interleukin-1 family. IL-1 consists of two distinct forms: IL1 alpha and IL1 beta that recognize the same cell surface receptors but are distinct proteins with approximately 25% amino acid sequence identity. IL1 alpha is constitutively produced by epithelial cells and plays an essential role in maintenance of skin barrier function. Upon stimulation, a wide variety of cells including osteoblasts, monocytes, macrophages can be induced to express IL1 alpha. IL1 alpha possesses a wide range of metabolic, physiological, haematopoietic activities, and is critically involved in the regulation of the immune responses and inflammatory responses.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité