Human recombinant IL1F10 (fromE. coli)
Fournisseur: ProSci Inc.
91-060
PRSI91-060EA 852 EUR InStock
PRSI91-060
Human recombinant IL1F10 (fromE. coli)
Protéines et Peptides
Human Interleukin 1 Family Member 10 (IL-1F10) is thought to participate in a network of Interleukin 1 cytokine family members to regulate adapted and innate immune responses. IL-1F10 was expressed in fetal skin, spleen and tonsil, mostly in the basal epithelia of skin and in proliferating B-cells of the tonsil. IL-1F10 binds soluble IL-1 receptor type 1 and may be implicated in regulating adapted and innate immune responses. Two alternatively spliced transcript variants encoding the same protein have been reported.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité