Human recombinant BDNF (from E. coli)
Fournisseur: ProSci Inc.
91-062
PRSI91-062EA 852 EUR InStock
PRSI91-062
Human recombinant BDNF (from E. coli)
Protéines et Peptides
Brain-Derived Neurotrophic Factor (BDNF) is a member of the neurotrophin family. Along with other structurally related neurotrophic factors NGF, NT-3 and NT-4, BDNF binds with high affinity to the TrkB kinase receptor. It also binds with the LNGFR (for low-affinity nerve growth factor receptor, also known as p75). BDNF promotes the survival, growth and differentiation of neurons. It serves as a major regulator of synaptic transmission and plasticity at adult synapses in many regions of the CNS. BDNF expression is altered in neurodegenerative disorders such as Parkinson's and Alzheimer's disease.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité