Human recombinant pro-BDNF (from E. coli)
Fournisseur: ProSci Inc.
91-063
PRSI91-063EA 654 EUR InStock
PRSI91-063
Human recombinant pro-BDNF (from E. coli)
Protéines et Peptides
The precursor form of Brain-Derived Neurotrophic Factor (pro-BDNF) interacts preferentially with the pan-neurotrophin receptor p75 (p75NTR) and vps10p domain-containing receptor sortilin and induces neuronal apoptosis, whereas mature BDNF selectively binds with high affinity to the TrkB kinase receptor and promotes the survival, growth and differentiation of neurons. As proneurotrophins and mature neurotrophins elicit opposite biological effects, Pro-BDNF cleavage in the neuronal system is regulated in a specific and cell-context dependent manner. Pro-BDNF plays important role in negative regulation of neurotrophic actions in the brain.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité