Human recombinant ProNGF (from E. coli)
Fournisseur: ProSci Inc.
91-064
PRSI91-064EA 654 EUR InStock
PRSI91-064
Human recombinant ProNGF (from E. coli)
Protéines et Peptides
The precursor form of the nerve growth factor (proNGF) like its mature form is characterized by the cystin knot motif consisting of three cystine bridges, whereas proneurotrophins and mature neurotrophins elicit opposite biological effects. ProNGF functions preferentially via the complex of pan-neurotrophin receptor p75 (p75NTR) and vps10p domain-containing receptor sortilin inducing neuronal apoptosis and contributing to age- and disease-related neurodegeneration.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité