Human recombinant VEGFA (from cells)
Fournisseur: ProSci Inc.
91-069
PRSI91-069EA 852 EUR InStock
PRSI91-069
Human recombinant VEGFA (from cells)
Protéines et Peptides
Human Vascular endothelial growth factor (VEGF), also known as VEGF-A and vascular permeability factor (VPF), belongs to the platelet-derived growth factor family of cysteine-knot growth factors. It is a potent activator in vasculogenesis and angiogenesis both physiologically and pathologically. VEGF-A has 8 differently spliced isoforms, of which VEGF165 is the most abundant one. VEGF165 is a disulfide-linked homodimer consisting of two glycosylated 165 amino acid polypeptide chains. VEGF stimulates the cellular response through binding to tyrosine kinase receptors VEGFR1 and VEGFR2 on the cell surface. It is widely accepted that VEGFR2 mediate almost all of the known cellular responses to VEGF while the function of VEGFR1 is less defined and is thought to modulate the VEGFR2 signaling.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité