Human recombinant Cystatin A (from E. coli)
Fournisseur: ProSci Inc.
91-071
PRSI91-071EA 549 EUR InStock
PRSI91-071
Human recombinant Cystatin A (from E. coli)
Protéines et Peptides
Human Cystatin A (CSTA) is a member of family 1 of the cystatin superfamily, which is characterized by lacking of disulphide bonds an carbohydrates. Cystatin A is an intracellular inhibitor regulating the activities of cysteine proteases of the papain family such as Cathepsins B, H and L. Cystatin A is also implicated in a number of disease states. Due to altered proteolytic state in cancer progression, Cystatin A may play a role in the proteolytic pathways.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité