Human recombinant CNTF (from E. coli)
Fournisseur: ProSci Inc.
91-081
PRSI91-081EA 734 EUR InStock
PRSI91-081
Human recombinant CNTF (from E. coli)
Protéines et Peptides
Ciliary Neurotrophic Factor (CNTF) is a potent survival factor for neurons and oligodendrocytes. CNTF has also been shown to prevent the degeneration of motor axons after axotomy. CNTF is highly conserved across species and exhibits cross-species activities. Human and rat CNTF share approximately 83% homology in their protein sequence. CNTF is structurally related to IL6, IL11, LIF and OSM. All of these four helix bundle cytokines share gp130 as a signal transducing subunit in their receptor complexes. CNTF, like FGF acidic, FGF basic, and PD-ECGF (platelet-derived endothelial cell growth factor), does not possess a signal sequence that would allow secretion of the factor by classical secretion pathways. The mechanism underlying the release of CNTF is unknown.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité