Human recombinant beta-Arrestin 1 (from E. coli)
Fournisseur: ProSci Inc.
91-085
PRSI91-085EA 654 EUR InStock
PRSI91-085
Human recombinant beta-Arrestin 1 (from E. coli)
Protéines et Peptides
beta -Arrestin-1 (ARRB1) is a cytoplasmic protein that belongs to the arrestin family. ARRB1 is expressed at high levels in peripheral blood leukocytes and the central nervous system. ARRB1 regulates agonist-mediated G-protein coupled receptor (GPCR) signaling by mediating both receptor desensitization and resensitization processes. ARRB1 acts as a cofactor in the beta-adrenergic receptor kinase (BARK) mediated desensitization of beta-adrenergic receptors. ARRB1 is believed to play a major role in regulating receptor-mediated immune functions. ARRB1 is involved in Toll-like receptor and IL-1 receptor signaling through the interaction with TRAF6.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité