Human recombinant BCL-2 Related A1 (from E. coli)
Fournisseur: ProSci Inc.
91-086
PRSI91-086EA 567 EUR InStock
PRSI91-086
Human recombinant BCL-2 Related A1 (from E. coli)
Protéines et Peptides
Bcl-2-Related Protein A1 (BCL2A1) is a member of of the BCL-2 protein family which act as anti- and pro-apoptotic regulators that are involved in a wide variety of cellular activities such as embryonic development, homeostasis and tumorigenesis. BCL2A1 is a cytoplasm protein and interacts directly with BAK1, BID, BMF and BBC3. BCL2A1 is induced by phorbol ester and inflammatory cytokines, such as TNF or IL1B/interleukin-1 beta, but not by growth factors. BCL2A1 is invovled in the response of hemopoietic cells to external signals and in maintaining endothelial survival during infection.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité