Human recombinant Carbonic Anhydrase 13 (from E. coli)
Fournisseur: ProSci Inc.
91-091
PRSI91-091EA 469 EUR InStock
PRSI91-091
Human recombinant Carbonic Anhydrase 13 (from E. coli)
Enzymes
Carbonic Anhydrase 13 (CA13) belongs to the carbonic anhydrase family which can catalyses the reversible hydration recation of carbon dioxide. Carbonic anhydrases participate in many biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. CA13 is a cytosolic enzyme and is widely expressed in human, such as thymus, small intestine, spleen, prostate, ovary, colon and testis, indicating that it may play a key role in several organs. CA13 is inhibited by acetazolamide.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité