Human recombinant Carbonic Anhydrase 4 (from E. coli)
Fournisseur: ProSci Inc.
91-092
PRSI91-092EA 789 EUR InStock
PRSI91-092
Human recombinant Carbonic Anhydrase 4 (from E. coli)
Enzymes
Carbonic Anhydrase 4 (CA4) belongs to the alpha-carbonic anhydrase family. Alpha-carbonic anhydrase is a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. Carbonic anhydrase 4 is a glycosylphosphatidyl-inositol-anchored membrane isozyme expressed on the luminal surfaces of pulmonary (and certain other) capillaries and proximal renal tubules. Carbonic anhydrase 4 may stimulate the sodium/bicarbonate transporter activity of SLC4A4 that acts in pH homeostasis. It may have a role in inherited renal abnormalities of bicarbonate transport. Furthermore, Carbonic anhydrase 4 is essential for acid overload removal from the retina and retina epithelium and acid release in the choriocapillaris.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité