Human Recombinant CCL26 (from E. coli)
Fournisseur: ProSci Inc.
91-096
PRSI91-096EA 734 EUR InStock
PRSI91-096
Human Recombinant CCL26 (from E. coli)
Protéines et Peptides
Chemokine Ligand 26 protein (CCL26) is a novel small cytokine belonging to the CC chemokine family which is involved in immunoregulatory and inflammatory processes. CCL26 is constitutively expressed in thymus, but only transiently expressed in phytohemagglutinin-stimulated peripheral blood mononuclear cells. It specifically binds and induces chemotaxis in T cells and elicits its effects by interacting with the chemokine receptor CCR4. CCL26, along with Eotaxin-1 and Eotaxin-2, selectively activates the CC chemokine receptor 3 (CCR3). The Eotaxin-3-CCR3 interaction may play an important role in allergic diseases such as atopic dermatitis and bronchial asthma. The full-length cDNA for CCL26 encodes a protein of 94 amino acids with a putative signal peptide of either 23 or 26 amino acid residues. Both the 71 and 68 amino acid residue variants of recombinant CCL26 demonstrate equal potency in inducing chemotaxis of a human CCR3-transfected cell line. Unlike most other CC chemokines, CCL26 maps to human chromosome 7q11.2, within 40 kilobases of the Eotaxin-2 loci. CCL26 and Eotaxin-2 are unique in that they are the only chemokines identified to date that map to chromosome 7.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité