E. coli recombinant lon protease (from E. coli)
Fournisseur: ProSci Inc.
91-099
PRSI91-099EA 0 EUR InStock
PRSI91-099
E. coli recombinant lon protease (from E. coli)
Enzymes
Lon Protease, is a member of the Lon protease family. They are found in archaea, bacteria and eukaryotes. Lon protease is ATP-dependent serine protease that mediates the selective degradation of mutant and abnormal proteins as well as certain short-lived regulatory proteins, including some antitoxins. It required for cellular homeostasis and for survival from DNA damage and developmental changes induced by stress. It degrades polypeptides processively to yield small peptide fragments that are 5 to 10 amino acids long and binds to DNA in a double-stranded, site-specific manner. Endogenous substrates include the regulatory proteins RcsA and SulA, the transcriptional activator SoxS, and UmuD. Its overproduction specifically inhibits translation through at least two different pathways, one of them being the YoeB-YefM toxin-antitoxin system.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité