Human recombinant CXCL12 (from E. coli)
Fournisseur: ProSci Inc.
91-105
PRSI91-105EA 852 EUR InStock
PRSI91-105
Human recombinant CXCL12 (from E. coli)
Protéines et Peptides
Stromal Cell-Derived Factor-1 (SDF-1) is a chemokine member of the intercrine family. SDF1 is expressed as five isoforms that differ only in the C terminal tail. SDF1 alpha and SDF1 beta are identical except for the four residues present in the C-terminus of SDF1 beta but absent from SDF1 alpha. SDF1 isoforms interact with CXCR4 and CXCR7 receptors on the cell surface, and can also bind syndecan4. SDF1 is known to influence lymphopoiesis, regulate patterning and cell number of neural progenitors, and promote angiogenesis. It also enhances the survival of myeloid progenitor cells.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité