Human recombinant beta-Defensin 1 (from E. coli)
Fournisseur: ProSci Inc.
91-108
PRSI91-108EA 654 EUR InStock
PRSI91-108
Human recombinant beta-Defensin 1 (from E. coli)
Protéines et Peptides
beta -Defensin 1 (DEFB1) is a member of the beta -defensin family, which is highly expressed by epithelial cells. beta -defensins are expressed as the C-terminal portion of precursors and are released by proteolytic cleavage of a signal peptide. beta -defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds. beta -defensin 1 is an antimicrobial peptide implicated in the resistance of epithelial surfaces to microbial colonization. Defects in beta -Defensin-1 contribute to asthma diagnosis, with apparent gender-specific effects in human. beta -defensin 1 may also play a role in the pathogenesis of severe sepsis. In addition, beta -defensin 1 is associated with induction profiles in gingival keratinocytes.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité