Human recombinant beta-Defensin 4 (from E. coli)
Fournisseur: ProSci Inc.
91-109
PRSI91-109EA 654 EUR InStock
PRSI91-109
Human recombinant beta-Defensin 4 (from E. coli)
Protéines et Peptides
Defensins are cationic peptides. It is an important ingredient of the innate immune system. beta -defensins are expressed on some leukocytes and epithelial surfaces. Four human beta -Defensins have been identified to date: BD-1, BD-2, BD-3 and BD-4. beta -defensins contain a six-cysteine motif, they forms three intra-molecular disulfide bonds. beta -defensins are also chemoattractant towards immature dendritic cells and memory T cells. The beta -defensin proteins are expressed as the C-terminal portion of precursors; they are released by proteolytic cleavage of a signal sequence.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité