Human recombinant Esterase D (from E. coli)
Fournisseur: ProSci Inc.
91-115
PRSI91-115EA 654 EUR InStock
PRSI91-115
Human recombinant Esterase D (from E. coli)
Enzymes
Human Esterase D is a serine hydrolase that is involved in the detoxification of formaldehyde. Esterase D plays a part in a variety of substrates, including O-acetylated sialic acids, which may involves in the recycling of sialic acids. Esterase D can be used as a genetic marker for retinoblastoma and Wilson’s disease.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité