Human recombinant FABP1 (from E. coli)
Fournisseur: ProSci Inc.
91-116
PRSI91-116EA 584 EUR InStock
PRSI91-116
Human recombinant FABP1 (from E. coli)
Protéines et Peptides
Human Fatty Acid-Binding Protein 1 (FABP1) is a cytoplasm protein, which belongs to the calycin superfamily and Fatty-acid binding protein (FABP) family. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids. FABP1 forms a beta-barrel structure that accommodates hydrophobic ligands in its interior. FABP1 can bind free fatty acids and their coenzyme A derivatives, bilirubin, and some other small molecules in the cytoplasm, so it can be involved in intracellular lipid transport.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité