Human recombinant FABP5 (from E. coli)
Fournisseur: ProSci Inc.
91-120
PRSI91-120EA 654 EUR InStock
PRSI91-120
Human recombinant FABP5 (from E. coli)
Protéines et Peptides
Fatty acid-binding protein 5 (FABP5) is a cytoplasm protein that belongs to the fatty-acid binding protein (FABP) family of calycin superfamily. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids. FABP5 can be expressed in keratinocytes, and is highly expressed in psoriatic skin. FABP5 has been shown to be involved in keratinocyte differentiation. FABP5 has high specificity for fatty acids, the highest affinity for C18 chain length. FABP5 can decrease the chain length or introduce double bonds to reduce the affinity.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité