Human Recombinant GOT1 (from E. coli)
Fournisseur: ProSci Inc.
91-127
PRSI91-127EA 654 EUR InStock
PRSI91-127
Human Recombinant GOT1 (from E. coli)
Protéines et Peptides
Glutamate Oxaloacetate Transaminase 1 (GOT1) is a cytoplasmic protein. GOT1 belongs to the class-I pyridoxal-phosphate-dependent aminotransferase family. GOT1 is a pyridoxal phosphate-dependent enzyme that exists in cytoplasmic and mitochondrial forms. GOT1 plays a key role in amino acid metabolism and the urea and tricarboxylic acid cycles. GOT1 involves in L-methionine salvage from methylthioadenosine, aspartate catabolic process, cellular response to insulin stimulus, polyamine metabolic process, and glucocorticoid stimulus.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité