Human recombinant LACTB2 (from E. coli)
Fournisseur: ProSci Inc.
91-131
PRSI91-131EA 654 EUR InStock
PRSI91-131
Human recombinant LACTB2 (from E. coli)
Protéines et Peptides
beta -Lactamase-like Protein 2 (LACTB2) is a number of the metallo-beta-lactamase superfamily.LACTB2 also belongs to the Glyoxalase II family. LACTB2 is 288 amino acids long with 8 zinc-binding domains. The LACTB2 gene is expressed at high levels and annotates structural defects or features in 4 cDNA clones. LACTB2 proteins are expected to have hydrolase activity and metal ion-binding functions. LACTB2 protein is found to localize in mitochondrion. Other functions of LACTB2 is yet unknown.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité