Human Recombinant NPPB (from E. coli)
Fournisseur: ProSci Inc.
91-136
PRSI91-136EA 584 EUR InStock
PRSI91-136
Human Recombinant NPPB (from E. coli)
Protéines et Peptides
Human Natriuretic peptides B acts as a cardiac hormone; it is associated with many biological actions, such as diuresis, natriuresis, vasorelaxation, which inhibits the secretion of rennin and aldosterone. It acts as a paracrine antifibrotic factor in the heart. Natriuretic peptides B can help restore the body balance of salt and water, improves the heart function. Natriuretic peptides B binds and stimulates the cGMP production of the NPR1 receptor and binds the clearance receptor NPR3.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité