Human recombinant PP1MG (from E. coli)
Fournisseur: ProSci Inc.
91-140
PRSI91-140EA 584 EUR InStock
PRSI91-140
Human recombinant PP1MG (from E. coli)
Protéines et Peptides
Protein Phosphatase 1G (PP1MG) is a cytoplasmic protein that belongs to the PP2C family. PPM1G is widely expressed; it is abundant in testis, skeletal muscle, and heart. PPM1G is a negative regulator of cell stress response pathways. PPM1G is responsible for the dephosphorylation of Pre-mRNA splicing factors, an important factor for the formation of functional spliceosome. PPM1G also plays a role in regulating cell cycle progression.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité