Human recombinant PPP1R14A (from E. coli)
Fournisseur: ProSci Inc.
91-142
PRSI91-142EA 584 EUR InStock
PRSI91-142
Human recombinant PPP1R14A (from E. coli)
Protéines et Peptides
Protein Phosphatase 1 Regulatory Subunit 14A (PPP1R14A) belongs to the PP1 inhibitor family. PPP1R14A is mapped to chromosome 19q13.13-q13.2. PPP1R14A binds directly to protein kinase C and casein kinase I. Meantime, PPP1R14A is a phosphorylation-dependent inhibitor of smooth muscle myosin phosphatase. PPP1R14A is the inhibitor of PPP1CA. When phosphorylated, PPP1R14A has over 1000-fold higher inhibitory activity, creating a molecular switch for regulating the phosphorylation status of PPP1CA substrates and smooth muscle contraction. In addition, inhibition of PPP1R14A also enhances contraction of smooth muscle in the absence of increment of intracellular Ca2+ concentration.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité