Human recombinant PPP1R1A (from E. coli)
Fournisseur: ProSci Inc.
91-143
PRSI91-143EA 584 EUR InStock
PRSI91-143
Human recombinant PPP1R1A (from E. coli)
Protéines et Peptides
Protein Phosphatase 1 Regulatory Subunit 1A (PPP1R1A) is an inhibitor of protein-phosphatase 1. PPP1R1A is a cellular regulator of eIF2 alpha phosphorylation. In hormonal control of glycogen metabolism, IPP-1 protein plays important function. Hormones can elevate intracellular cAMP level and elevate IPP-1 activity. PPP1R1A activation caused cAMP increase , cAMP control over proteins that are not directly phosphorylated by PKA following a rise in intracellular calcium. IPP-1 is inactivated by calcineurin (PP2B). Multiple domains in IPP-1 target cellular PP1 complexes.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité