Human Recombinant Gankyrin (from E. coli)
Fournisseur: ProSci Inc.
91-146
PRSI91-146EA 584 EUR InStock
PRSI91-146
Human Recombinant Gankyrin (from E. coli)
Protéines et Peptides
Gankyrin is a multicatalytic proteinase oncoprotein consists of 7 ankyrin repeats. Gankyrin overexpressed in most hepatocellular carcinomas. Gankyrin is involved in theregulation of the phosphorylation of the retinoblastoma protein by CDK4 to enhance the ubiquitinylation of p53 by MDM2. Gankyrin is also involved in progression of esophageal squamous cell carcinoma. Gankyrin plays an oncogenic role especially in early stages of human epatocarcinogenesis.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité