Human Recombinant 40S Ribosomal S19 (from E. coli)
Fournisseur: ProSci Inc.
91-150
PRSI91-150EA 584 EUR InStock
PRSI91-150
Human Recombinant 40S Ribosomal S19 (from E. coli)
Protéines et Peptides
40S Ribosomal Protein S19 (RPS19) is a ribosomal protein that Belongs to the ribosomal protein S19e family. RPS19 is located in the nucleoli, and higher level expression is seen in colon carcinoma tissue than normal colon tissue. It required for pre-rRNA processing and maturation of 40S ribosomal subunits. RPS19 plays a role in many biological processes, such as endocrine pancreas development, erythrocyte differentiation, mRNA metabolic process. Defects in RPS19 are the cause of Diamond-Blackfan anemia type 1 (DBA1), which is a form of Diamond-Blackfan anemia, a congenital non-regenerative hypoplastic anemia that usually presents early in infancy. Diamond-Blackfan anemia is characterized by a moderate to severe macrocytic anemia, erythroblastopenia, and an increased risk of malignancy.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité