Human recombinant Sentrin-specific protease 8 (from E. coli)
Fournisseur: ProSci Inc.
91-152
PRSI91-152EA 343 EUR InStock
PRSI91-152
Human recombinant Sentrin-specific protease 8 (from E. coli)
Enzymes
Sentrin-Specific Protease 8 (SENP8) mediates the reversible covalent modification of proteins by NEDD8. SENP8 catalyses the full-length NEDD8 to generate its mature form and deconjugation of NEDD8 from targeted proteins such as CUL2 , CUL4A in vivo, or p53. but it does not show activity against ubiquitin or SUMO proteins.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité