Human recombinant TNF beta (from E. coli)
Fournisseur: ProSci Inc.
91-161
PRSI91-161EA 734 EUR InStock
PRSI91-161
Human recombinant TNF beta (from E. coli)
Protéines et Peptides
Tumor Necrosis Factor beta (TNF- beta ) is a secreted protein belonging to the tumor necrosis factor family. TNF- beta binds to TNFRSF1A/TNFR1, TNFRSF1B/TNFBR and TNFRSF14/HVEM in homotrimeric form, binds to TNFRSF3/LTBR in heterotrimeric form with LTB. TNF- beta forms heterotrimers with lymphotoxin-beta, which anchors TNF- beta to the cell surface. TNF- beta mediates the inflammatory, immunostimulatory, and antiviral response, involves in the formation of second lymphoid organs during development, has a role in apoptosis. TNF- beta is produced by lymphocytes and cytotoxic for a variety of tumor cells in vitro and in vivo.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité