Human recombinant Thioredoxin 2 (from E. coli)
Fournisseur: ProSci Inc.
91-162
PRSI91-162EA 654 EUR InStock
PRSI91-162
Human recombinant Thioredoxin 2 (from E. coli)
Protéines et Peptides
Thioredoxin-2 (TXN2) is a mitochondrial member of the thioredoxin family. Thioredoxin-2 is extensively expressed in adult and fetal tissues. Thioredoxin-2 contains an N-terminal 59 amino acid transit peptide, which is cleaved before translocating to mitochondria. Mitochondrial thioredoxin play important roles in the regulation of the mitochondrial membrane potential and in protection against oxidant-induced apoptosis. Thioredoxin-2 could be involved in the resistance to anti-tumor agents and possesses a dithiol-reducing activity. In addition, Thioredoxin-2 is important at low oxidative stress conditions.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité