Human recombinant UBE2K (from E. coli)
Fournisseur: ProSci Inc.
91-166
PRSI91-166EA 343 EUR InStock
PRSI91-166
Human recombinant UBE2K (from E. coli)
Protéines et Peptides
Ubiquitin-Conjugating Enzyme E2 K (UBE2K) belongs to the E2 Ubiquitin-Conjugating Enzyme family. UBE2K is highly expressed in the brain, with highest levels found in cortex and striatum, and at lower levels in cerebellum and brainstem. UBE2K may mediate foam cell formation by the suppression of apoptosis of lipid-bearing macrophages through ubiquitination and subsequence degradation of p53/TP53. UBE2K is associated with the selective degradation of short-lived and abnormal proteins, such as the endoplasmic reticulum-associated degradation (ERAD) of misfolded lumenal proteins. In addition, UBE2K is involved in Alzheimer's disease, Huntington's disease and antigen processing through its interaction with huntingtin, and MHC-heavy chain proteins.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité