Human recombinant UBE2T (from E. coli)
Fournisseur: ProSci Inc.
91-167
PRSI91-167EA 343 EUR InStock
PRSI91-167
Human recombinant UBE2T (from E. coli)
Protéines et Peptides
Ubiquitin-Conjugating Enzyme E2 T (UBE2T) is a ligase that belongs to the Ubiquitin-Conjugating Enzyme family. UBE2T accepts the ATP-dependent ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro, UBE2T is able to catalyze polyubiquitination using all 7 ubiquitin Lys residues, but may prefer 'Lys-11'-, 'Lys-27'-, 'Lys-48'- and 'Lys-63'-linked polyubiquitination. UBE2T is an important factor of the Faconi anemia pathway of DNA damage repair and, upon self-inactivation, may negatively regulate the Faconi pathway.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité