Human recombinant UCH-L1 (from E. coli)
Fournisseur: ProSci Inc.
91-168
PRSI91-168EA 409 EUR InStock
PRSI91-168
Human recombinant UCH-L1 (from E. coli)
Protéines et Peptides
Ubiquitin Carboxyl-Terminal Hydrolase Isozyme L1 (UCHL1) belongs to the Peptidase C12 family. UCHL1 is specifically expressed in the neurons and in cells of the diffuse neuroendocrine system. UCHL1 is a component of the ubiquitin system, which has a fundamental role in regulating various biological activities. UCHL1 is a thiol protease that recognizes and hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin. UCHL1 also binds to free monoubiquitin and may prevent its degradation in lysosomes. The homodimer of UCHL1 may have ATP-independent ubiquitin ligase activity.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité