Human recombinant UCH-L3 (from E. coli)
Fournisseur: ProSci Inc.
91-169
PRSI91-169EA 813 EUR InStock
PRSI91-169
Human recombinant UCH-L3 (from E. coli)
Protéines et Peptides
Ubiquitin Carboxyl-Terminal Hydrolases (UCHs) are a family of cysteine hydrolases. They catalyze the hydrolysis of amides, thioesters and esters, peptide and isopeptide bonds formed by the C-terminal Gly of ubiquitin. Up regulation of UCHL3 is associated with uterine cervical neoplasms. UCHL3 is implicated in age related cognitive disorders. UCHL3 also promotes adipogenesis and insulin signaling. In mice, UCHL3 knockout have been shown to be resistant to diet-induced obesity.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité