Human recombinant Serpin A12 (from E. coli)
Fournisseur: ProSci Inc.
91-172
PRSI91-172EA 584 EUR InStock
PRSI91-172
Human recombinant Serpin A12 (from E. coli)
Protéines et Peptides
Vaspin (Visceral Adipose-Specific SERPIN) is a newly described adipokine. Vaspin has three beta -sheets, nine alpha-helices, and one central loop; the structure is part of the set of distinctive features that are descriptive of Serpin family members. Vaspin is also a unique insulin sensitizing adipocytokine in obesity. A recent publication indicates that Vaspin mRNA expression in visceral fat is positively correlated with BMI and percent of body fat. and could be associated with parameters of obesity, insulin resistance, and glucose metabolism. These findings suggest a potential clinical use for Vaspin in ameliorating certain aberrations seen in the obesity metabolic syndrome.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité