Human recombinant ARC (from E. coli)
Fournisseur: ProSci Inc.
91-177
PRSI91-177EA 654 EUR InStock
PRSI91-177
Human recombinant ARC (from E. coli)
Protéines et Peptides
Nucleolar Protein 3 is encoded by NOL3 gene; multiple transcript variants encoding different isoforms have been found for this gene. So far, Nucleolar protein 3 has show to have two Isoforms. Isoform 1 may be involved in RNA splicing.Isoform 2 may inhibit apoptosis.It has been shown to down-regulate the enzyme activities of caspase 2, caspase 8 and tumor protein p53.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité