Human Recombinant EIF4E-Binding 1 (from E. coli)
Fournisseur: ProSci Inc.
91-180
PRSI91-180EA 654 EUR InStock
PRSI91-180
Human Recombinant EIF4E-Binding 1 (from E. coli)
Protéines et Peptides
Eukaryotic Translation Initiation Factor 4E-Binding Protein 1 (4EBP1) is a number of the eIF4E-binding protein family. 4EBP1 regulates eIF4E activity by preventing its assembly into the eIF4F complex. 4EBP1 mediates the regulation of protein translation by hormones, growth factors and other stimuli that signal through the MAP kinase and mTORC1 pathways. Non-phosphorylated 4EBP1 competes with EIF4G1/EIF4G3 to interact with EIF4E. 4EBP1 is phosphorylated in response to various signals including insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation. 4EBP1 has a role in progression of breast neoplasms through cell signaling.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité