Human recombinant Angiogenin (from E. coli)
Fournisseur: ProSci Inc.
91-183
PRSI91-183EA 549 EUR InStock
PRSI91-183
Human recombinant Angiogenin (from E. coli)
Protéines et Peptides
Angiogenin belongs to the pancreatic ribonuclease family. Angiogenin is primarily expressed in the liver. It may act as a tRNA-specific ribonuclease that abolishes protein synthesis by specifically hydrolyzing cellular tRNAs. Angiogenin is a potent stimulator of new blood vessel formation. And Angiogenin is endocytosed and translocated to the nucleus by binding to actin on the surface of endothelial cells. Angiogenic activity is regulated by interaction with RNH1 in vivo. In addition, Angiogenin is associated with susceptibility to amyotrophic lateral sclerosis, which is a degenerative disorder of motor neurons in the cortex, brain stem and spinal cord.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité