Human recombinant Annexin A13 (from E. coli)
Fournisseur: ProSci Inc.
91-184
PRSI91-184EA 654 EUR InStock
PRSI91-184
Human recombinant Annexin A13 (from E. coli)
Protéines et Peptides
Annexin A13 (ANXA13) belongs to the annexin family which plays a role in phospholipase inhibition, cytoskeletal interactions, intracellular signal transduction pathways and regulation of cellular growth. ANXA13 contains four annexin repeats and a pair of annexin repeats may form one binding site for calcium and phospholipid. ANXA13 is highly expressed in intestinal and kidney epithelial cells. The specific function of ANXA13 has not yet been determined; however it is associated with the plasma membrane of undifferentiated, proliferating crypt epithelial cells as well as differentiated villus enterocytes.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité