Human recombinant Annexin A5 (from E. coli)
Fournisseur: ProSci Inc.
91-186
PRSI91-186EA 654 EUR InStock
PRSI91-186
Human recombinant Annexin A5 (from E. coli)
Protéines et Peptides
Annexin A5 (ANXA5) is a member of the annexin family of calcium-dependent phospholipid binding proteins. ANXA5 is an anticoagulant protein by acting as an indirect inhibitor of the thromboplastin-specific complex. ANXA5 is also a protein kinase C and phospholipase A2 inhibitor. It participate in inflammation, cellular signal transduction, growth and differentiation. ANXA5 binding to phosphatidylserine and sulfatide to regulates coagulability in the blood stream. It also protects sinsuoidal endothelial cells from ischemia reperfusion damage. ANXA5 is important for normal CFTR chloride channel activity.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité