Human recombinant Annexin A6 (from E. coli)
Fournisseur: ProSci Inc.
91-187
PRSI91-187EA 549 EUR InStock
PRSI91-187
Human recombinant Annexin A6 (from E. coli)
Protéines et Peptides
Annexin A6 (ANAX6) belongs to a family of calcium-dependent membrane and phospholipid binding proteins. Annexin A6 is a secreted protein and locates on the cell surface. Annexin A6 contains 8 annexin repeats separated by linking sequences of variable lengths. A pair of annexin repeats may form one binding site for calcium and phospholipid. ANXA6 is involved in the regulation of the release of Ca2+ from intracellular stores and may be associated with CD21. In addition, ANXA6 has been implicated in mediating the endosome aggregation and vesicle fusion in secreting epithelia during exocytosis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité