Human recombinant Annexin A7 (from E. coli)
Fournisseur: ProSci Inc.
91-188
PRSI91-188EA 584 EUR InStock
PRSI91-188
Human recombinant Annexin A7 (from E. coli)
Protéines et Peptides
Annexin A7 (ANXA7) is a member of the annexin family of calcium-dependent phospholipid binding proteins. Annexin A7 has a unique, highly hydrophobic N-terminal domain and a conserved C-terminal region. The C-terminal region is composed of alternating hydrophobic and hydrophilic segments. Annexin A7 is a calcium/phospholipid-binding protein with diverse properties including voltage-sensitive calcium channel activity and promotes membrane fusion and is also involved in exocytosis.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité