Human recombinant CLIC5 (from E. coli)
Fournisseur: ProSci Inc.
91-192
PRSI91-192EA 852 EUR InStock
PRSI91-192
Human recombinant CLIC5 (from E. coli)
Protéines et Peptides
Chloride Intracellular Channel Protein 5 (CLIC5) is a single-pass membrane protein which belongs to the chloride channel CLIC family. It contains one GST C-terminal domain. Chloride intracellular channels are involved in chloride ion transport within various subcellular compartments. CLIC5 can insert into membranes and form selective ion channels regulated by actin that may transport chloride ions. It may play a role in the regulation of transepithelial ion absorption and secretion. CLIC5 specifically associates with the cytoskeleton of placenta microvilli. CLIC5 is required for the development and/or maintenance of the proper glomerular endothelial cell and podocyte architecture.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité