Human recombinant Cyclophilin E (from E. coli)
Fournisseur: ProSci Inc.
91-196
PRSI91-196EA 884 EUR InStock
PRSI91-196
Human recombinant Cyclophilin E (from E. coli)
Protéines et Peptides
Peptidyl-prolyl cis-trans isomerase E, also known as Cyclophilin E, Cyclophilin-33, Rotamase E, CYP33, PPIE, is an enzyme which belongs to the cyclophilin-type PPIase family of PPIase E subfamily. PPIE found in all the examined tissues including heart, brain, placenta, lung, liver, skeletal muscle, kidney and pancreas. PPIE contains one PPIase cyclophilin-type domain and one RRM (RNA recognition motif) domain. PPIE accelerates the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides. PPIE combines RNA-binding and PPIase activities. It may be involved in muscle- and brain-specific processes and pre-mRNA splici
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité